missing translation for 'onlineSavingsMsg'
Learn More

4-n-Heptylbenzoic acid, 99+%

Product Code. 11475167
Change view
Click to view available options
Quantity:
5 g
25 g
Unit Size:
25g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11475167 5 g 5g
11485167 25 g 25g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11475167 Supplier Thermo Scientific Alfa Aesar Supplier No. B22589.06
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 38350-87-7 | C14H20O2 | 220.312 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 38350-87-7
Molecular Formula C14H20O2
Molecular Weight (g/mol) 220.312
MDL Number MFCD00009722
InChI Key VSUKEWPHURLYTK-UHFFFAOYSA-N
Synonym benzoic acid, 4-heptyl, 4-n-heptylbenzoic acid, 4-heptylbenzoicacid, 4-heptylbenzoate, p-heptylbenzoic acid, acmc-209izm, 4-n-heptyl-benzoic acid, 4-heptylbenzoic acid
PubChem CID 170036
IUPAC Name 4-heptylbenzoic acid
SMILES CCCCCCCC1=CC=C(C=C1)C(=O)O

Specifications

Melting Point 101°C to 102°C
Quantity 5 g
Beilstein 2047135
Formula Weight 220.31
Percent Purity ≥99%
Chemical Name or Material 4-n-Heptylbenzoic acid

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.