missing translation for 'onlineSavingsMsg'
Learn More

4-n-Dodecyloxybenzaldehyde, 98%

Product Code. 11382766
Change view
Click to view available options
Quantity:
1 g
5 g
Unit Size:
1g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11382766 1 g 1g
11392766 5 g 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11382766 Supplier Thermo Scientific Alfa Aesar Supplier No. L08889.03
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 24083-19-0 | C19H30O2 | 290.447 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 24083-19-0
Molecular Formula C19H30O2
Molecular Weight (g/mol) 290.447
MDL Number MFCD00043523
InChI Key ZBEGLEYBWGNZJA-UHFFFAOYSA-N
Synonym p-dodecyloxybenzaldehyde, 4-dodecyloxy benzaldehyde, 4-dodecyloxybenzaldehyde, 4-n-dodecyloxybenzaldehyde, p-n-dodecoxy benzaldehyde, p-dodecyloxy benzaldehyde, 4-dodecyloxy-benzaldehyde, p-lauryloxy benzaldehyde, 1-4-formylphenoxy dodecane
PubChem CID 141069
IUPAC Name 4-dodecoxybenzaldehyde
SMILES CCCCCCCCCCCCOC1=CC=C(C=C1)C=O

Specifications

Melting Point 26°C to 29°C
Boiling Point 165°C (0.005 mmHg)
Quantity 1 g
Beilstein 2535031
Sensitivity Air Sensitive
Formula Weight 290.45
Percent Purity 98%
Chemical Name or Material 4-n-Dodecyloxybenzaldehyde

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.