missing translation for 'onlineSavingsMsg'
Learn More

4-n-Dodecylbenzoic acid, 98%

Product Code. 15494637
Change view
Click to view available options
Quantity:
1 g
5 g
Unit Size:
1g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
15494637 1 g 1g
15404647 5 g 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 15494637 Supplier Thermo Scientific Alfa Aesar Supplier No. H31983.03
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 21021-55-6 | C19H30O2 | 290.447 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 21021-55-6
Molecular Formula C19H30O2
Molecular Weight (g/mol) 290.447
MDL Number MFCD04972235
InChI Key JCHCIOJFCCQEKZ-UHFFFAOYSA-N
Synonym p-dodecylbenzoic acid, benzoic acid, 4-dodecyl, p-n-dodecyl benzoic acid, benzoicacid,4-dodecyl, 4-n-dodecylbenzoic acid
PubChem CID 413311
IUPAC Name 4-dodecylbenzoic acid
SMILES CCCCCCCCCCCCC1=CC=C(C=C1)C(=O)O

Especificaciones

Quantity 1 g
Formula Weight 290.45
Percent Purity 98%
Chemical Name or Material 4-n-Dodecylbenzoic acid

RUO – Research Use Only

Título del producto
Seleccione un problema

Al hacer clic en Enviar, acepta que Fisher Scientific se ponga en contacto con usted en relación con los comentarios que ha proporcionado en este formulario. No compartiremos su información para ningún otro fin. Toda la información de contacto proporcionada se mantendrá de acuerdo con nuestra Política de Privacidad. Política de privacidad.