missing translation for 'onlineSavingsMsg'
Learn More

4-n-Decylbenzoyl chloride, 98%

Product Code. 11433457
Change view
Click to view available options
Quantity:
1 g
5 g
25 g
Unit Size:
1g
25g
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11433457 1 g 1g
11443457 5 g 5g
11453457 25 g 25g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 11433457 Supplier Thermo Scientific Alfa Aesar Supplier No. A17234.03
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

CAS: 54256-43-8 | C17H25ClO | 280.836 g/mol

Employed as a pharmaceutical and chemical intermediate.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Employed as a pharmaceutical and chemical intermediate.

Solubility
Reacts with water.

Notes
Moisture Sensitive. Store away from oxidizing agents, water/moisture. Store under inert gas.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 54256-43-8
Molecular Formula C17H25ClO
Molecular Weight (g/mol) 280.836
MDL Number MFCD00041724
InChI Key HEWLDHSFOQXCSI-UHFFFAOYSA-N
PubChem CID 104714
IUPAC Name 4-decylbenzoyl chloride
SMILES CCCCCCCCCCC1=CC=C(C=C1)C(=O)Cl

Specifications

Boiling Point 171°C to 173°C (2 mmHg)
Flash Point >110°C (230°F)
Quantity 1 g
UN Number UN3265
Sensitivity Moisture sensitive
Synonym 4-n-decylbenzoyl chloride, benzoyl chloride, 4-decyl, p-decylbenzoyl chloride, 4-decylbenzoylchloride, p-n-decylbenzoyl chloride, benzoyl chloride,4-decyl, 4-n-decyl benzoyl chloride, 4-decylbenzoic acid chloride
Solubility Information Reacts with water.
Formula Weight 280.84
Percent Purity 98%
Chemical Name or Material 4-n-Decylbenzoyl chloride

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.