missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Thermo Scientific™ 4-(N-Cyclopentylcarbamoyl)benzeneboronic acid pinacol ester, 95%
Click to view available options
:
1 g
Specifications
Specifications
| CAS | 933987-10-1 |
| Molecular Formula | C{1}{8}H{2}{6}BNO{3} |
| MDL Number | MFCD16660857 |
| Quantity | 1 g |
| Synonym | N-Cyclopentyl-4-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)benzamide; |
| Formula Weight | 315.22 |
| Percent Purity | 95% |
| Physical Form | Solid |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction