missing translation for 'onlineSavingsMsg'
Learn More

4-n-Butylbenzylamine, 98%

Product Code. 11436006
Change view
Click to view available options
Quantity:
5 g
25 g
100 g
Unit Size:
100g
25g
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11436006 25 g 25g
11426006 5 g 5g
11446006 100 g 100g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 11436006 Supplier Thermo Scientific Alfa Aesar Supplier No. B22316.14
 Restricted Item
Estimated Shipment: 07-12-2026
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

CAS: 57802-79-6 | C11H17N | 163.264 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 57802-79-6
Molecular Formula C11H17N
Molecular Weight (g/mol) 163.264
MDL Number MFCD02258855
InChI Key IBVGSPOHLFKLHM-UHFFFAOYSA-N
Synonym 4-butylbenzylamine, 4-n-butylbenzylamine, 4-butylphenyl methanamine, 4-butylphenyl methylamine, 1-4-butylphenyl methanamine, p-n-butylbenzylamin, 4-butyl-benzylamine, 4-butylbenzenemethanamine, benzenemethanamine, 4-butyl
PubChem CID 4029871
IUPAC Name (4-butylphenyl)methanamine
SMILES CCCCC1=CC=C(C=C1)CN

Specifications

Boiling Point 89°C to 91°C (1 mmHg)
Quantity 25 g
UN Number UN2735
Sensitivity Air Sensitive
Formula Weight 163.26
Percent Purity 98%
Chemical Name or Material 4-n-Butylbenzylamine

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.