missing translation for 'onlineSavingsMsg'
Learn More

4-n-Butylbenzoic acid, 99%

Product Code. 11426746
Change view
Click to view available options
Quantity:
5 g
25 g
100 g
Unit Size:
100g
25g
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11426746 25 g 25g
11416746 5 g 5g
11436746 100 g 100g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 11426746 Supplier Thermo Scientific Alfa Aesar Supplier No. B23891.14
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 20651-71-2 | C11H14O2 | 178.231 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 20651-71-2
Molecular Formula C11H14O2
Molecular Weight (g/mol) 178.231
MDL Number MFCD00002571
InChI Key JFKUBRAOUZEZSL-UHFFFAOYSA-N
Synonym 4-n-butylbenzoic acid, benzoic acid, 4-butyl, 4-butyl-benzoic acid, p-butylbenzoic acid, p-n-butylbenzoic acid, p-butyl benzoic acid, 4-butylbenzoicacid, p-n-butyl benzoic acid, acmc-209fcp, 4-n-butyl-benzoic acid
PubChem CID 88631
IUPAC Name 4-butylbenzoic acid
SMILES CCCCC1=CC=C(C=C1)C(=O)O

Specifications

Melting Point 100°C to 113°C
Quantity 25 g
Formula Weight 178.23
Percent Purity 99%
Chemical Name or Material 4-n-Butylbenzoic acid

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.