missing translation for 'onlineSavingsMsg'
Learn More

4-n-Butylaniline, 97%, Thermo Scientific™

Product Code. 11394689
Change view
Click to view available options
Quantity:
25 g
100 g
500 g
Unit Size:
100g
25g
500g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11394689 25 g 25g
11304699 100 g 100g
15493277 500 g 500g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 11394689 Supplier Thermo Scientific Alfa Aesar Supplier No. B25234.14
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 104-13-2
Molecular Formula C10H15N
Molecular Weight (g/mol) 149.237
MDL Number MFCD00007925
InChI Key OGIQUQKNJJTLSZ-UHFFFAOYSA-N
Synonym 4-n-butylaniline, benzenamine, 4-butyl, p-n-butylaniline, p-butylaniline, 4-butylbenzenamine, p-aminobutylbenzene, p-butylaminobenzene, 1-amino-4-butylbenzene, p-butyl aniline, 4-butylphenylamine
PubChem CID 7694
IUPAC Name 4-butylaniline
SMILES CCCCC1=CC=C(C=C1)N

Specifications

Melting Point -14°C
Density 0.942
Boiling Point 250°C
Flash Point 101°C (213°F)
Refractive Index 1.535
Quantity 25 g
Beilstein 1447268
Sensitivity Air Sensitive
Formula Weight 149.23
Percent Purity 97%
Chemical Name or Material 4-n-Butylaniline
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.