missing translation for 'onlineSavingsMsg'
Learn More

4-n-Butoxyphenol, 98%, Thermo Scientific Chemicals

Catalog Number 11499972
Click to view available options
Quantity:
5 g
25 g
100 g
This item is not returnable. View return policy
122-94-1
C10H14O2
166.22
MFCD00002336
MBGGFXOXUIDRJD-UHFFFAOYSA-N
phenol, 4-butoxy, p-butoxyphenol, 4-n-butoxyphenol, phenol, p-butoxy, hydroquinone monobutyl ether, unii-8188dbs68j, 4-normal-butoxyphenol, 4-butoxy-pheno, 4-butoxy-phenol, 4-n-butyloxyphenol
31233
CHEBI:34392
4-butoxyphenol
CCCCOC1=CC=C(O)C=C1
This item is not returnable. View return policy
122-94-1
C10H14O2
166.22
MFCD00002336
MBGGFXOXUIDRJD-UHFFFAOYSA-N
phenol, 4-butoxy, p-butoxyphenol, 4-n-butoxyphenol, phenol, p-butoxy, hydroquinone monobutyl ether, unii-8188dbs68j, 4-normal-butoxyphenol, 4-butoxy-pheno, 4-butoxy-phenol, 4-n-butyloxyphenol
31233
CHEBI:34392
4-butoxyphenol
CCCCOC1=CC=C(O)C=C1

It is an intermediate of liquid crystals.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
It is an intermediate of liquid crystals.

Solubility
Soluble in acetone and ethanol.

Notes
Store in cool place. Keep away from oxidizing agents.
TRUSTED_SUSTAINABILITY
Melting Point 63°C to 66°C
Quantity 5 g
Solubility Information Soluble in acetone and ethanol.
Formula Weight 166.22
Percent Purity 98%
Chemical Name or Material 4-n-Butoxyphenol

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'