missing translation for 'onlineSavingsMsg'
Learn More

4-n-Butoxyphenol, 98%

Product Code. 11419982
Change view
Click to view available options
Quantity:
5 g
25 g
100 g
Unit Size:
100g
25g
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11419982 100 g 100g
11499972 5 g 5g
11409982 25 g 25g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 11419982 Supplier Thermo Scientific Alfa Aesar Supplier No. A12076.22
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 122-94-1 | C10H14O2 | 166.22 g/mol

It is an intermediate of liquid crystals.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
It is an intermediate of liquid crystals.

Solubility
Soluble in acetone and ethanol.

Notes
Store in cool place. Keep away from oxidizing agents.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 122-94-1
Molecular Formula C10H14O2
Molecular Weight (g/mol) 166.22
MDL Number MFCD00002336
InChI Key MBGGFXOXUIDRJD-UHFFFAOYSA-N
Synonym phenol, 4-butoxy, p-butoxyphenol, 4-n-butoxyphenol, phenol, p-butoxy, hydroquinone monobutyl ether, unii-8188dbs68j, 4-normal-butoxyphenol, 4-butoxy-pheno, 4-butoxy-phenol, 4-n-butyloxyphenol
PubChem CID 31233
ChEBI CHEBI:34392
IUPAC Name 4-butoxyphenol
SMILES CCCCOC1=CC=C(O)C=C1

Specifications

Melting Point 63°C to 66°C
Quantity 100 g
Solubility Information Soluble in acetone and ethanol.
Formula Weight 166.22
Percent Purity 98%
Chemical Name or Material 4-n-Butoxyphenol

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.