missing translation for 'onlineSavingsMsg'
Learn More

4-n-Butoxybenzoyl chloride, 98%

Product Code. 11494773
Change view
Click to view available options
Quantity:
5 g
25 g
100 g
Unit Size:
100g
25g
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11494773 25 g 25g
11484773 5 g 5g
11404783 100 g 100g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 11494773 Supplier Thermo Scientific Alfa Aesar Supplier No. A11658.14
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 33863-86-4 | C11H13ClO2 | 212.673 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 33863-86-4
Molecular Formula C11H13ClO2
Molecular Weight (g/mol) 212.673
MDL Number MFCD00000689
InChI Key KMGCTFHTBKBITO-UHFFFAOYSA-N
PubChem CID 118561
IUPAC Name 4-butoxybenzoyl chloride
SMILES CCCCOC1=CC=C(C=C1)C(=O)Cl

Specifications

Density 1.121
Boiling Point 161°C to 162°C (9 mmHg)
Flash Point >110°C (230°F)
Refractive Index 1.5495
Quantity 25 g
UN Number UN3265
Beilstein 957203
Sensitivity Moisture sensitive
Synonym 4-n-butoxybenzoyl chloride, 4-butoxy-benzoyl chloride, benzoyl chloride, 4-butoxy, p-butoxybenzoyl chloride, 4-n-butoxybenzoylchloride, 4-butyloxy benzoyl chloride, benzoyl chloride,4-butoxy, 4-n-butoxy benzoyl chloride, 4-n-butyloxy benzoyl chloride
Formula Weight 212.68
Percent Purity 98%
Chemical Name or Material 4-n-Butoxybenzoyl chloride
Show More Show Less

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.