missing translation for 'onlineSavingsMsg'
Learn More

4-n-Butoxybenzonitrile, 97%, Thermo Scientific™

Product Code. 11427897
Change view
Click to view available options
Quantity:
5 g
25 g
Unit Size:
25g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11427897 5 g 5g
11437897 25 g 25g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11427897 Supplier Thermo Scientific Alfa Aesar Supplier No. A16570.06
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 5203-14-5
Molecular Formula C11H13NO
Molecular Weight (g/mol) 175.231
MDL Number MFCD00043482
InChI Key RRGQINKVTNAIBB-UHFFFAOYSA-N
Synonym p-butoxybenzonitrile, 4-n-butoxybenzonitrile, benzonitrile, 4-butoxy, 4-n-butoxy benzonitrile, 4-butoxy-benzonitrile, 4-butoxybenzenecarbonitrile, pubchem7926, acmc-209kxf, 4-butoxybenzonitrile
PubChem CID 138435
IUPAC Name 4-butoxybenzonitrile
SMILES CCCCOC1=CC=C(C=C1)C#N

Specifications

Density 0.999
Boiling Point 242°C to 244°C
Flash Point >110°C (230°F)
Odor Characteristic
Refractive Index 1.5255
Quantity 5 g
Beilstein 2834664
Formula Weight 175.23
Percent Purity 97%
Chemical Name or Material 4-n-Butoxybenzonitrile
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.