missing translation for 'onlineSavingsMsg'
Learn More

4-n-Butoxybenzoic Acid, TRC

Product Code. 30328881
Change view
Click to view available options
Quantity:
500 mg
Unit Size:
500mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30328881 500 mg 500mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30328881 Supplier Toronto Research Chemicals Supplier No. TRCB692328500MG
 Restricted Item
Estimated Shipment: 02-12-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material 4-n-Butoxybenzoic Acid
CAS 1498-96-0
Molecular Formula C11H14O3
Quantity 500 mg
Synonym p-(Butyloxy)benzoic Acid, p-(n-Butoxy)benzoic Acid, p-(n-Butyloxy)benzoic Acid, p-Butoxybenzoic Acid, 4-Butoxybenzoic Acid, 4-(Butyloxy)benzoic Acid
InChI Formula InChI=1S/C11H14O3/c1-2-3-8-14-10-6-4-9(5-7-10)11(12)13/h4-7H,2-3,8H2,1H3,(H,12,13)
SMILES CCCCOc1ccc(cc1)C(=O)O
IUPAC Name 4-butoxybenzoic acid
Molecular Weight (g/mol) 194.23
Formula Weight 194.0943
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.