missing translation for 'onlineSavingsMsg'
Learn More

4-Morpholinylsulfur trifluoride, 95%, Thermo Scientific Chemicals

Catalog Number 11359685
Click to view available options
Quantity:
1 g
5 g
25 g
This item is not returnable. View return policy
51010-74-3
C4H8F3NOS
175.169
MFCD00037057
UFXIRMVZNARBDL-UHFFFAOYSA-N
morpholinosulfur trifluoride, morpholinosulfurtrifluoride, morph-dast, 4-trifluoro-$l^ 4-sulfanyl morpholine, 4-trifluoro-??-sulfanyl morpholine, morpholinosulphur trifluoride, morpho-dast, acmc-209krh, trifluoro morpholin-4-yl, morpholinotrifluorosulfur iv
2733258
trifluoro(morpholin-4-yl)-$l^{4}-sulfane
C1COCCN1S(F)(F)F
This item is not returnable. View return policy
51010-74-3
C4H8F3NOS
175.169
MFCD00037057
UFXIRMVZNARBDL-UHFFFAOYSA-N
morpholinosulfur trifluoride, morpholinosulfurtrifluoride, morph-dast, 4-trifluoro-$l^ 4-sulfanyl morpholine, 4-trifluoro-??-sulfanyl morpholine, morpholinosulphur trifluoride, morpho-dast, acmc-209krh, trifluoro morpholin-4-yl, morpholinotrifluorosulfur iv
2733258
trifluoro(morpholin-4-yl)-$l^{4}-sulfane
C1COCCN1S(F)(F)F

4-Morpholinylsulfur trifluoride is incorporated in the fluoronation of trihexanols. It is used as a nucleophilic fluorinating agent with greater thermal stability. It is also used in the preparation of proline analogs and in the preparation of difluoropiperidines.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
4-Morpholinylsulfur trifluoride is incorporated in the fluoronation of trihexanols. It is used as a nucleophilic fluorinating agent with greater thermal stability. It is also used in the preparation of proline analogs and in the preparation of difluoropiperidines.

Solubility
Reacts with water.

Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Incompatible with bases and oxidizing and bases. Store under dry inert gas. It is sensitive to moisture.
TRUSTED_SUSTAINABILITY
Density 1.436
Boiling Point 41°C to 42°C (0.5 mmHg)
Flash Point 76°C (168°F)
Refractive Index 1.4545
Quantity 1 g
UN Number UN3265
Beilstein 606224
Sensitivity Moisture sensitive; May form a precipitate
Solubility Information Reacts with water.
Formula Weight 175.17
Percent Purity 95%
Chemical Name or Material 4-Morpholinylsulfur trifluoride
Show More Show Less

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'