Learn More
4-Morpholinylsulfur trifluoride, 95%
Description
4-Morpholinylsulfur trifluoride is incorporated in the fluoronation of trihexanols. It is used as a nucleophilic fluorinating agent with greater thermal stability. It is also used in the preparation of proline analogs and in the preparation of difluoropiperidines.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
4-Morpholinylsulfur trifluoride is incorporated in the fluoronation of trihexanols. It is used as a nucleophilic fluorinating agent with greater thermal stability. It is also used in the preparation of proline analogs and in the preparation of difluoropiperidines.
Solubility
Reacts with water.
Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Incompatible with bases and oxidizing and bases. Store under dry inert gas. It is sensitive to moisture.
Specifications
Specifications
| Density | 1.436 |
| Boiling Point | 41°C to 42°C (0.5 mmHg) |
| Flash Point | 76°C (168°F) |
| Refractive Index | 1.4545 |
| Quantity | 5 g |
| UN Number | UN3265 |
| Beilstein | 606224 |
| Sensitivity | Moisture sensitive; May form a precipitate |
| Solubility Information | Reacts with water. |
| Formula Weight | 175.17 |
| Show More |
RUO – Research Use Only
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.