missing translation for 'onlineSavingsMsg'
Learn More

4-(Morpholinosulfonyl)aniline, 97%, Thermo Scientific™

Catalog Number p-2700860
Click to view available options
Quantity:
1 g
5 g
10 g
This item is not returnable. View return policy
21626-70-0
C10H14N2O3S
242.293
MFCD00277213
FTKHPQFFQRKOJC-UHFFFAOYSA-N
4-morpholinosulfonyl aniline, 4-morpholin-4-ylsulfonyl aniline, 4-morpholine-4-sulfonyl aniline, compound 82 208, 4-morpholine-4-sulfonyl-phenylamine, 4-4-morpholinylsulfonyl aniline, compound 82/208, morpholine, 4-sulfanilyl, sulfanilic acid, morpholide, 4-aminophenyl morpholino sulfone
210794
4-morpholin-4-ylsulfonylaniline
C1COCCN1S(=O)(=O)C2=CC=C(C=C2)N
This item is not returnable. View return policy
21626-70-0
C10H14N2O3S
242.293
MFCD00277213
FTKHPQFFQRKOJC-UHFFFAOYSA-N
4-morpholinosulfonyl aniline, 4-morpholin-4-ylsulfonyl aniline, 4-morpholine-4-sulfonyl aniline, compound 82 208, 4-morpholine-4-sulfonyl-phenylamine, 4-4-morpholinylsulfonyl aniline, compound 82/208, morpholine, 4-sulfanilyl, sulfanilic acid, morpholide, 4-aminophenyl morpholino sulfone
210794
4-morpholin-4-ylsulfonylaniline
C1COCCN1S(=O)(=O)C2=CC=C(C=C2)N
CAS Min % 97.0
Assay Percent Range 97%
Packaging Amber Glass Bottle
Quantity 1 g
Formula Weight 242.3
Percent Purity 97%
Chemical Name or Material 4-(Morpholinosulfonyl)aniline
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'