missing translation for 'onlineSavingsMsg'
Learn More

4-(Morpholinomethyl)benzoic Acid, TRC

Product Code. 30482394
Change view
Click to view available options
Quantity:
1 g
Unit Size:
1g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30482394 1 g 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30482394 Supplier Toronto Research Chemicals Supplier No. TRCM7251801G
 Restricted Item
Estimated Shipment: 25-08-2026
Add to basket
Add to basket
This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material 4-(Morpholinomethyl)benzoic Acid
CAS 62642-62-0
Molecular Formula C10 H8 Cl N O2 S
Quantity 1 g
Synonym 4-(4-Morpholinylmethyl)benzoic Acid, 4-(Morpholin-4-ylmethyl)benzoic Acid
InChI Formula InChI=1S/C10H8ClNO2S/c1-7-5-6-12-10-8(7)3-2-4-9(10)15(11,13)14/h2-6H,1H3
SMILES Cc1ccnc2c(cccc12)S(=O)(=O)Cl
IUPAC Name 4-methylquinoline-8-sulfonyl chloride
Molecular Weight (g/mol) 241.694
Formula Weight 240.996
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.