missing translation for 'onlineSavingsMsg'
Learn More

4-(Morpholinomethyl)benzaldehyde, 97%, Thermo Scientific™

Product Code. 10444962
Change view
Click to view available options
Quantity:
250 mg
1 g
10 g
Packaging:
Amber Glass Bottle
Unit Size:
10g
1g
250mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging unitSize
10444962 1 g Amber Glass Bottle 1g
10046242 250 mg Amber Glass Bottle 250mg
10613455 10 g Amber Glass Bottle 10g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 10444962 Supplier Thermo Scientific Maybridge Supplier No. CC29304DA
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Chemical Identifiers

CAS 82413-63-6
Molecular Formula C12H15NO2
Molecular Weight (g/mol) 205.257
MDL Number MFCD05865107
InChI Key KMAHWHPUXGNVBN-UHFFFAOYSA-N
Synonym 4-morpholinomethyl benzaldehyde, 4-morpholin-4-ylmethyl benzaldehyde, 4-morpholinomethyl-benzaldehyde, 4-4-morpholinylmethyl benzaldehyde, 4-morpholin-4-ylmethyl-benzaldehyde, 4-4-carboxaldehydebenzyl morpholine, benzaldehyde,4-4-morpholinylmethyl
PubChem CID 2795498
IUPAC Name 4-(morpholin-4-ylmethyl)benzaldehyde
SMILES C1COCCN1CC2=CC=C(C=C2)C=O

Specifications

CAS Min % 94.5
CAS Max % 100.0
Assay Percent Range 95%
Packaging Amber Glass Bottle
Quantity 1 g
Formula Weight 205.26
Percent Purity 95%
Chemical Name or Material 4-(Morpholinomethyl)benzaldehyde
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.