missing translation for 'onlineSavingsMsg'
Learn More

4-(Morpholinomethyl)aniline, 95+%, Thermo Scientific™

Product Code. 10357304
Change view
Click to view available options
Quantity:
250 mg
1 g
5 g
Packaging:
Amber Glass Bottle
Unit Size:
1g
250mg
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging unitSize
10357304 5 g Amber Glass Bottle 5g
10550423 250 mg Amber Glass Bottle 250mg
10517863 1 g Amber Glass Bottle 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 10357304 Supplier Thermo Scientific Maybridge Supplier No. CC29314DE
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Chemical Identifiers

CAS 51013-67-3
Molecular Formula C11H16N2O
Molecular Weight (g/mol) 192.262
InChI Key WNYFVEFUHMDIRQ-UHFFFAOYSA-N
Synonym 4-morpholinomethyl aniline, 4-morpholin-4-ylmethyl aniline, 4-morpholin-4-ylmethylphenylamine, 4-morpholin-4-ylmethyl-phenylamine, benzenamine, 4-4-morpholinylmethyl, 4-4-morpholinylmethyl aniline, 4-morpholin-4-ylmethyl-aniline, 4-morpholin-4-ylmethyl phenylamine, acmc-209krj, 4-morpholinomethyl-aniline
PubChem CID 776851
IUPAC Name 4-(morpholin-4-ylmethyl)aniline
SMILES C1COCCN1CC2=CC=C(C=C2)N

Specifications

Assay Percent Range 95%
Packaging Amber Glass Bottle
Quantity 5 g
Formula Weight 192.26
Percent Purity 95%
Chemical Name or Material 4-(Morpholinomethyl)aniline
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.