missing translation for 'onlineSavingsMsg'
Learn More

4-(morpholinocarbonyl)phenylboronic acid, 97%, Maybridge

Product Code. 10422474
Change view
Click to view available options
Quantity:
1 g
Packaging:
Amber Glass Bottle
Unit Size:
1g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging unitSize
10422474 1 g Amber Glass Bottle 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 10422474 Supplier Thermo Scientific Maybridge Supplier No. MAY00021DA
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

Specifications

CAS 656239-38-2
CAS Min % 97.0
Assay Percent Range 97%
Packaging Amber Glass Bottle
Molecular Formula C17H24BNO4
MDL Number MFCD05863913
Quantity 1 g
Synonym morpholino 4-4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl phenyl methanone, 4-morpholine-4-carbonyl phenylboronic acid pinacol ester, 4-morpholinocarbonyl phenylboronic acid, pinacol ester, 4-4-4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl benzoyl morpholine, 4-morpholine carboxamidophenylboronic acid, pinacol ester, morpholine, 4-4-4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl benzoyl, morpholin-4-yl-4-4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl-phenyl-methanone, morpholin-4-yl-4-4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl phenyl methanone, 4-4-morpholinylcarbonyl benzeneboronic acid pinacol ester, 4-morpholine-4-carbonyl phenyl boronic acid pinacol ester
InChI Key ZOPBEWUNCNLYSD-UHFFFAOYSA-N
SMILES CC1(C)OB(OC1(C)C)C1=CC=C(C=C1)C(=O)N1CCOCC1
IUPAC Name 4-[4-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)benzoyl]morpholine
Molecular Weight (g/mol) 317.19
PubChem CID 2760141
Formula Weight 317.19
Percent Purity 97%
Chemical Name or Material 4-(Morpholinocarbonyl)phenylboronic acid, pinacol ester
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.