missing translation for 'onlineSavingsMsg'
Learn More

4-Morpholinobenzenecarbothioamide, 97%, Thermo Scientific™

Product Code. 10451274
Click to view available options
Quantity:
250 mg
1 g
5 g
This item is not returnable. View return policy
This item is not returnable. View return policy

Chemical Identifiers

CAS 519056-60-1
Molecular Formula C11H14N2OS
Molecular Weight (g/mol) 222.31
MDL Number MFCD04115379
InChI Key KOPFTYFPHHZQCH-UHFFFAOYSA-N
Synonym 4-morpholinobenzenecarbothioamide, 4-morpholinobenzothioamide, 4-morpholin-4-yl benzenecarbothioamide, 4-morpholin-4'-y lthiobenzamide, amino 4-morpholin-4-ylphenyl methane-1-thione
PubChem CID 2795360
IUPAC Name 4-morpholin-4-ylbenzenecarbothioamide
SMILES NC(=S)C1=CC=C(C=C1)N1CCOCC1

Specifications

CAS Min % 96.5
CAS Max % 100.0
Assay Percent Range 97%
Packaging Amber glass bottle
Quantity 1 g
Formula Weight 222.31
Percent Purity 97%
Chemical Name or Material 4-Morpholinobenzenecarbothioamide
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'