missing translation for 'onlineSavingsMsg'
Learn More

4-Morpholinoacetophenone, 99%, Thermo Scientific Chemicals

Product Code. 10215770 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
5 g
50 g
Packaging:
Glass bottle
Unit Size:
50g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging unitSize
10215770 50 g Glass bottle 50g
10647122 5 g Glass bottle 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 10215770 Supplier Thermo Scientific Acros Supplier No. 152900500
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 39910-98-0
Molecular Formula C12H15NO2
Molecular Weight (g/mol) 205.26
MDL Number MFCD00006168
InChI Key AKQWEDMTPCAESO-UHFFFAOYSA-N
Synonym 4'-morpholinoacetophenone, 1-4-morpholinophenyl ethanone, 4-morpholinoacetophenone, acetophenone, 4'-morpholino, ethanone, 1-4-4-morpholinyl phenyl, p-morpholinoacetophenone, 1-4-morpholin-4-yl phenyl ethan-1-one, p-morpholino acetophenone, 4;-morpholinoacetophenone, 1-4-morpholin-4-ylphenyl ethanone
PubChem CID 38362
IUPAC Name 1-(4-morpholin-4-ylphenyl)ethanone
SMILES CC(=O)C1=CC=C(C=C1)N2CCOCC2

Specifications

CAS Min % 98.5
CAS Max % 100.0
Melting Point 94°C to 98°C
Color Yellow to Brown
Infrared Spectrum Authentic
Assay Percent Range 98.5% min. (GC)
Packaging Glass bottle
Quantity 50 g
Formula Weight 205.26
Percent Purity 99%
Physical Form Crystalline Powder
Chemical Name or Material 4-Morpholinoacetophenone, 99%
Show More Show Less
compliance-icons
Product Identifier
  • 4-Morpholinoacetophenone
Signal Word
  • Warning
Hazard Category
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
  • H315-Causes skin irritation.
Precautionary Statement
  • P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.