missing translation for 'onlineSavingsMsg'
Learn More

(4-Morpholin-4-yl-phenyl)-methanol, 95+%, Thermo Scientific™

Product Code. 10161062
Change view
Click to view available options
Quantity:
250 mg
1 g
10 g
Packaging:
Amber Glass Bottle
Unit Size:
10g
1g
250mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging unitSize
10161062 1 g Amber Glass Bottle 1g
10693203 250 mg Amber Glass Bottle 250mg
10441274 10 g Amber Glass Bottle 10g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 10161062 Supplier Thermo Scientific Maybridge Supplier No. CC17409DA
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Chemical Identifiers

CAS 280556-71-0
Molecular Formula C11H15NO2
Molecular Weight (g/mol) 193.25
MDL Number MFCD01057413
InChI Key KUAHZNZWSIDSTH-UHFFFAOYSA-N
Synonym 4-morpholin-4-yl-phenyl methanol, 4-morpholinobenzyl alcohol, 4-morpholin-4-yl phenyl methanol, 4-morpholinophenyl methanol, 4-morpholin-4-yl-phenyl-methanol, 4-morpholin-4-ylphenyl methanol, 4-morpholin-4-yl benzyl alcohol, 4-morpholin-4-ylphenyl methan-1-ol, 4-morpholinobenzylalcohol, 4-4-morpholinyl benzenemethanol
PubChem CID 2776456
SMILES OCC1=CC=C(C=C1)N1CCOCC1

Specifications

Assay Percent Range 95%
Packaging Amber Glass Bottle
Quantity 1 g
IUPAC Name (4-morpholin-4-ylphenyl)methanol
Formula Weight 193.24
Percent Purity 95%
Chemical Name or Material (4-Morpholin-4-yl-phenyl)-methanol
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.