missing translation for 'onlineSavingsMsg'
Learn More

4-Methylvaleric Acid (Isocaproic Acid), Mikromol™

Product Code. 30050595
Change view
Click to view available options
Quantity:
100 mg
Unit Size:
100mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30050595 100 mg 100mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30050595 Supplier Mikromol Supplier No. MM0863.10

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Discover Mikromol - your trusted source for high-quality pharmaceutical reference standards. Supporting accurate, compliant analysis with ISO 17034-accredited materials designed for confidence in every result.

4-Methylvaleric Acid (Isocaproic Acid)
TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material Isocaproic Acid
CAS 646-07-1
Analyte or Component Names Isocaproic Acid
Shipping Condition Room Temperature
Molecular Formula C6 H12 O2
Certifications/Compliance ISO/IEC 17025
Synonym 4-Methylpentanoic acid, Valeric acid, 4-methyl- (6CI,8CI), 4,4-Dimethylbutanoic acid, 4-Methylpentanoic acid, 4-Methylvaleric acid, Isobutylacetic acid, Isocaproic acid, Isohexanoic acid, Isohexoic acid, NSC 4126
InChI Formula InChI=1S/C6H12O2/c1-5(2)3-4-6(7)8/h5H,3-4H2,1-2H3,(H,7,8)
SMILES CC(C)CCC(=O)O
IUPAC Name 4-methylpentanoic acid
Molecular Weight (g/mol) 116.16
Quantity 100 mg
Formula Weight 116.0837 g/mol
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.