missing translation for 'onlineSavingsMsg'
Learn More

4-Methylvaleric Acid Ethyl Ester, TRC

Product Code. 30483187
Change view
Click to view available options
Quantity:
1 g
5 g
10 g
Unit Size:
10g
1g
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30483187 5 g 5g
30466978 1 g 1g
30465827 10 g 10g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 30483187 Supplier Toronto Research Chemicals Supplier No. TRCM3385005G
 Restricted Item

This item requires a price update prior to being ordered.

Request a Price for this item here.

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 25415-67-2
Molecular Formula C8H16O2
Molecular Weight (g/mol) 144.21
Synonym Ethyl 4-Methylpentanoate, Ethyl 4-Methylvalerate, Ethyl Isocaproate, Ethyl Isohexanoate, 4-Methylpentanoic Acid Ethyl Ester
IUPAC Name ethyl 4-methylpentanoate
SMILES O=C(OCC)CCC(C)C

Specifications

Quantity 5 g
InChI Formula InChI=1S/C8H16O2/c1-4-10-8(9)6-5-7(2)3/h7H,4-6H2,1-3H3
Formula Weight 144.115
Chemical Name or Material 4-Methylvaleric Acid Ethyl Ester
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.