missing translation for 'onlineSavingsMsg'
Learn More
Learn More
4-Methylvaleric acid, Dr. Ehrenstorfer
Specifications
Specifications
| Chemical Name or Material | Isocaproic Acid |
| CAS | 646-07-1 |
| Analyte or Component Names | Isocaproic Acid |
| Shipping Condition | Room Temperature |
| Molecular Formula | C6 H12 O2 |
| Certifications/Compliance | ISO/IEC 17025 |
| Synonym | 4-Methylpentanoic acid, Valeric acid, 4-methyl- (6CI,8CI), 4,4-Dimethylbutanoic acid, 4-Methylpentanoic acid, 4-Methylvaleric acid, Isobutylacetic acid, Isocaproic acid, Isohexanoic acid, Isohexoic acid, NSC 4126 |
| InChI Formula | InChI=1S/C6H12O2/c1-5(2)3-4-6(7)8/h5H,3-4H2,1-2H3,(H,7,8) |
| Solution Type | Neat |
| SMILES | CC(C)CCC(=O)O |
| Show More |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?