missing translation for 'onlineSavingsMsg'
Learn More

4-Methylvaleric acid, 99%

Product Code. 10399140 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
5 mL
25 mL
Packaging:
Glass bottle
Unit Size:
25mL
5mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging Unit Size
10399140 25 mL Glass bottle 25mL
10389140 5 mL Glass bottle 5mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 10399140 Supplier Thermo Scientific Acros Supplier No. 255510250
 Restricted Item
Estimated Shipment: 02-12-2026
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

CAS: 646-07-1 | C6H12O2 | 116.15 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 646-07-1
Molecular Formula C6H12O2
Molecular Weight (g/mol) 116.15
MDL Number MFCD00002803
InChI Key FGKJLKRYENPLQH-UHFFFAOYSA-N
Synonym 4-methylvaleric acid, isocaproic acid, isohexanoic acid, pentanoic acid, 4-methyl, isobutylacetic acid, 4-methyl-pentanoic acid, valeric acid, 4-methyl, 4-methyl valeric acid, isocaproate, 4,4-dimethylbutanoic acid
PubChem CID 12587
ChEBI CHEBI:74903
IUPAC Name 4-methylpentanoic acid
SMILES CC(C)CCC(=O)O

Specifications

CAS Min % 98.5
CAS Max % 100.0
Melting Point -35.0°C
Color Colorless to Yellow
Density 0.9220g/mL
Boiling Point 200.0°C
Flash Point 97°C
Infrared Spectrum Authentic
Assay Percent Range 98.5% min. (GC)
Packaging Glass bottle
Linear Formula (CH3)2CHCH2CH2COOH
Refractive Index 1.4136 to 1.4156
Quantity 25 mL
Specific Gravity 0.922
Solubility Information Solubility in water: insoluble
Formula Weight 116.15
Percent Purity 99%
Physical Form Liquid
Chemical Name or Material 4-Methylvaleric acid, 99%
Show More Show Less

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.