missing translation for 'onlineSavingsMsg'
Learn More

4-Methylumbelliferyl sulfate potassium salt, 98%

Product Code. 15487409
Change view
Click to view available options
Quantity:
500 mg
1 g
5 g
Unit Size:
1g
500mg
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
15487409 1 g 1g
15410517 500 mg 500mg
15497409 5 g 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 15487409 Supplier Thermo Scientific Alfa Aesar Supplier No. J66160.03
 Restricted Item
Estimated Shipment: 18-12-2026
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Substrate for sulfatase | CAS: 15220-11-8 | C10H7KO6S | 294.318 g/mol

4-Methylumbelliferyl sulfate potassium salt is a fluorometric substrate for the determination of sulfatase. This substrate has been used in the discovery of tyrosine sulfotransferase inhibitors, particularly in P-selectin glycoprotein ligand-1 binding. This compound is virtually non-fluorescent until cleavage by sulfatase enzymes, then emits blue fluorescence (450 nm) with excitation at 360 nm. Use of this compound in high-throughput fluorescence based screening led to the discovery of several potent beta-arylsulfotransferase IV (beta-AST-IV) inhibitors.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
4-Methylumbelliferyl sulfate potassium salt is a fluorometric substrate for the determination of sulfatase. This substrate has been used in the discovery of tyrosine sulfotransferase inhibitors, particularly in P-selectin glycoprotein ligand-1 binding. This compound is virtually non-fluorescent until cleavage by sulfatase enzymes, then emits blue fluorescence (450 nm) with excitation at 360 nm. Use of this compound in high-throughput fluorescence based screening led to the discovery of several potent beta-arylsulfotransferase IV (beta-AST-IV) inhibitors.

Solubility
Soluble in water at 5mg/ml

Notes
Excitation wavelength, 334 nm; emission wavelength, 370nm (pH 10.4). Reaction product: excitiation wavelength, 360nm; emission wavelength, 449nm
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 15220-11-8
Molecular Formula C10H7KO6S
Molecular Weight (g/mol) 294.318
MDL Number MFCD00016970
InChI Key CSOCSPXOODWGLJ-UHFFFAOYSA-M
Synonym 4-methylumbelliferyl sulfate potassium salt, 2h-1-benzopyran-2-one, 4-methyl-7-sulfooxy-, potassium salt, 4-methylumbelliferyl sulfate, potassium salt, 2h-1-benzopyran-2-one, 4-methyl-7-sulfooxy-, potassium salt 1:1, potassium 4-methyl-2-oxo-2h-chromen-7-yl sulfate, potassium 4-methyl-2-oxochromen-7-yl sulfate, c10h7o6s.k, potassium 4-methyl-2-oxo-2h-1-benzopyran-7-yl sulphate, potassium 4-methylumbelliferyl sulfate
PubChem CID 5044226
IUPAC Name potassium;(4-methyl-2-oxochromen-7-yl) sulfate
SMILES CC1=CC(=O)OC2=C1C=CC(=C2)OS(=O)(=O)[O-].[K+]

Specifications

Color White to Yellow
Quantity 1 g
Beilstein 3803203
Solubility Information Soluble in water at 5mg/ml
Formula Weight 294.32
Percent Purity 98%
Physical Form Powder
Chemical Name or Material 4-Methylumbelliferyl sulfate potassium salt

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.