missing translation for 'onlineSavingsMsg'
Learn More

4-Methylumbelliferyl Phosphate Bis-(2-amino- 2-methyl-1,3-propanediol) Salt, TRC

Product Code. 30483748
Change view
Click to view available options
Quantity:
100 mg
Unit Size:
100mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30483748 100 mg 100mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30483748 Supplier Toronto Research Chemicals Supplier No. TRCM337648100MG
 Restricted Item
Estimated Shipment: 13-08-2026
Add to basket
Add to basket
This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material 4-Methylumbelliferyl Phosphate Bis-(2-amino- 2-methyl-1,3-propanediol) Salt
CAS 107475-10-5
Molecular Formula C10H9O6P .2(C4H11NO2)
Quantity 100 mg
Synonym 2-Amino-2-methyl-1,3-Propanediol compd. with 4-Methyl-7-(phosphonooxy)-2H-1-benzopyran-2-one
InChI Formula InChI=1S/C11H15ClN2O2S/c12-8-5-6-11(10(7-8)14-13)17(15,16)9-3-1-2-4-9/h5-7,9,14H,1-4,13H2
SMILES NNc1cc(Cl)ccc1S(=O)(=O)C2CCCC2
IUPAC Name (5-chloro-2-cyclopentylsulfonylphenyl)hydrazine
Molecular Weight (g/mol) 466.4
Formula Weight 274.0543
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.