missing translation for 'onlineSavingsMsg'
Learn More

4-Methylumbelliferyl phosphate, 99%

Product Code. 15447319
Click to view available options
Quantity:
250 mg
1 g
5 g
Unit Size:
1g
250mg
5g
This item is not returnable. View return policy

Product Code. 15447319

Brand: Thermo Scientific Alfa Aesar J66091.06

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 3368-04-5 | C10H9O6P | 256.15 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 3368-04-5
MDL Number MFCD00016969

Specifications

Melting Point 214°C to 216°C
Color White
Molecular Formula C10H9O6P
Quantity 5 g
Beilstein 1687229
Synonym 4-methylumbelliferyl phosphate, 4-methyl-2-oxo-2h-chromen-7-yl dihydrogen phosphate, 4-mup, 2h-1-benzopyran-2-one, 4-methyl-7-phosphonooxy, phosphoric acid 4-methylumbelliferyl ester, 4-methylumbelliferylphosphate, free acid, 4-methyl-2-oxochromen-7-yl oxyphosphonic acid, 5,5-dinitrobarbutyric acid, 4-methylumbelliferone phosphate, 4-methylumbelliferyl phosphate, free acid
Solubility Information Soluble in water
InChI Key BCHIXGBGRHLSBE-UHFFFAOYSA-N
SMILES CC1=CC(=O)OC2=C1C=CC(=C2)OP(=O)(O)O
IUPAC Name (4-methyl-2-oxochromen-7-yl) dihydrogen phosphate
Molecular Weight (g/mol) 256.15
PubChem CID 65118
Formula Weight 256.15
Percent Purity 99%
Physical Form Powder
Chemical Name or Material 4-Methylumbelliferyl phosphate
Show More Show Less

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.