missing translation for 'onlineSavingsMsg'
Learn More

4-Methylumbelliferyl phosphate, 99%

Product Code. 15437319
Change view
Click to view available options
Quantity:
250 mg
1 g
5 g
Unit Size:
1g
250mg
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
15437319 1 g 1g
15457319 250 mg 250mg
15447319 5 g 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 15437319 Supplier Thermo Scientific Alfa Aesar Supplier No. J66091.03
 Restricted Item
In Stock
Add to basket
This item is not returnable. View return policy

CAS: 3368-04-5 | C10H9O6P | 256.15 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 3368-04-5
MDL Number MFCD00016969

Specifications

Melting Point 214°C to 216°C
Color White
Molecular Formula C12H11N2NaO3
Quantity 1 g
Beilstein 1687229
Synonym nalidixic acid sodium salt, baktogram, nalidixic acid sodium, sodium nalidixate, chemiurin, dixilina, nalidixate sodium anhydrous, nalidixate sodium, nalidixate sodium anhydrous usan, 1-ethyl-1,4-dihydro-7-methyl-4-oxo-1,8-naphthyridine-3-carboxylic acid sodium salt
Solubility Information Soluble in water
InChI Key ROKRAUFZFDQWLE-UHFFFAOYSA-M
SMILES CCN1C=C(C(=O)C2=C1N=C(C=C2)C)C(=O)[O-].[Na+]
IUPAC Name sodium;1-ethyl-7-methyl-4-oxo-1,8-naphthyridine-3-carboxylate
Molecular Weight (g/mol) 254.22
PubChem CID 3864541
Formula Weight 256.15
Percent Purity 99%
Physical Form Powder
Chemical Name or Material 4-Methylumbelliferyl phosphate
Show More Show Less

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.