missing translation for 'onlineSavingsMsg'
Learn More

MedChemExpress 4-Methylumbelliferyl phosphate

Product Code. 16424697
Encompass_Preferred
Change view
Click to view available options
Quantity:
50 mg
Unit Size:
50mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
16424697 50 mg 50mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 16424697 Supplier MedChemExpress Supplier No. HYD0994

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

MedChemExpress 4-Methylumbelliferyl phosphate (4-MUP) is a fluorogenic substrate of alkaline phosphatases.

Specifications

CAS 3368-04-5
Solubility H2O : 20.83 mg/mL (81.32 mM∣Need ultrasonic)
Synonym 4-MUP∣ MUP
Content And Storage -20°C, protect from light∣In solvent : -80°C, 6 months∣-20°C, 1 month (protect from light)
For Use With (Application) Neuroscience-Neuromodulation
Quantity 50 mg
Molecular Weight (g/mol) 256.15 g/mol
Product Type Dye Reagents
Purity or Quality Grade 99.88%
Shipping Condition Room temperature
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.