Learn More
Melford 4-Methylumbelliferyl-N-acetyl-B-D-glucosaminide
Product Code. p-200087284
Click to view available options
Quantity:
100 mg
1 g
5 g
Unit Size:
100mg
1g
5g
Product Code. 30018498
Brand: Melford M641000.1
Fluorescent substrate for N-Acetyl-B-D-glucosaminidase and B-N-Acetyl-D-hexosaminidase.
Fluorescent substrate for N-Acetyl-B-D-glucosaminidase and B-N-Acetyl-D-hexosaminidase. Ultrasensitive. Checked by actual enzymatic assay.
Specifications
Quantity 100 mg