Learn More
Melford 4-Methylumbelliferyl-α-L-arabinopyranoside
Product Code. 30018502
Change view
Click to view available options
Quantity:
100 mg
Unit Size:
100mg
Product Code. Quantity Unit Size
30018502 100 mg 100mg
1 options
Product Code. 30018502 Supplier Melford Supplier No. M641550.1
 Restricted Item
Estimated Shipment: 09-10-2026
Add to basket
Add to basket
missing translation for 'validMainframeMsg'
Fluorescent substrate for α-L-Arabinosidase.
Fluorescent substrate for α-L-Arabinosidase. 4-Methylumbelliferone < 50ppm.
Specifications
Quantity 100 mg