Learn More
Melford 4-Methylumbelliferyl-α-D-mannopyranoside
Product Code. p-200087266
Click to view available options
Quantity:
25 mg
100 mg
Unit Size:
100mg
25mg
Product Code. 30018528
Brand: Melford M666000.025
Fluorescent substrate for α-D-Mannosidase.
Fluorescent substrate for α-D-Mannosidase. 4-Methylumbelliferone: <50ppm.
Specifications
Quantity 25 mg