Learn More
Melford 4-Methylumbelliferyl-α-D-mannopyranoside
Product Code. 30018528
Change view
Click to view available options
Quantity:
25 mg
100 mg
Unit Size:
100mg
25mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30018528 25 mg 25mg
30018529 100 mg 100mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Product Code. 30018528 Supplier Melford Supplier No. M666000.025
 Restricted Item
Estimated Shipment: 28-07-2026
Add to basket
Add to basket
Fluorescent substrate for α-D-Mannosidase.
Fluorescent substrate for α-D-Mannosidase. 4-Methylumbelliferone: <50ppm.
Specifications
Quantity 25 mg