Learn More
Melford 4-Methylumbelliferyl-β-D-cellobiopyranoside
Product Code. p-200087282
Click to view available options
Quantity:
100 mg
500 mg
Unit Size:
100mg
500mg
Product Code. 30018505
Brand: Melford M646000.5
Fluorescent substrate for β-D-Cellobiopyranoside.
Fluorescent substrate for β-D-Cellobiopyranoside.
4-Methylumbelliferone < 50ppm.
Specifications
Quantity 500 mg