missing translation for 'onlineSavingsMsg'
Learn More

4-Methylumbelliferyl Beta-D-glucuronide Dihydrate, TRC

Product Code. 30478847
Change view
Click to view available options
Quantity:
500 mg
Unit Size:
500mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30478847 500 mg 500mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30478847 Supplier Toronto Research Chemicals Supplier No. TRCM849655500MG

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material 4-Methylumbelliferyl beta-D-glucuronide Dihydrate
Molecular Formula C16H16O9 . 2H 2O
Quantity 500 mg
Synonym Glucopyranosiduronic acid, 4-methyl-2-oxo-2H-1-benzopyran-7-yl, β-D- (8CI), 4-Methyl-2-oxo-2H-1-benzopyran-7-yl β-D-glucopyranosiduronic acid, 4-Methylumbelliferone glucuronide, 4-Methylumbelliferone β-D-glucuronide, 4-Methylumbelliferyl glucuronide, 4-Methylumbelliferyl β-D-glucuronide, 4-Methylumbelliferyl-β-D-glucuronic acid, Hymecromon-7-O-4-β-D-glucuronide, β-D-Glucopyranosiduronic acid, 4-methyl-2-oxo-2H-1-benzopyran-7-yl
InChI Formula InChI=1S/C16H16O9.2H2O/c1-6-4-10(17)24-9-5-7(2-3-8(6)9)23-16-13(20)11(18)12(19)14(25-16)15(21)22;;/h2-5,11-14,16,18-20H,1H3,(H,21,22);2*1H2/t11-,12-,13+,14-,16+;;/m0../s1
SMILES CC1=CC(OC2=C1C=CC(O[C@H]3[C@H](O)[C@@H](O)[C@H](O)[C@@H](C(O)=O)O3)=C2)=O.O.O
IUPAC Name (2S,3S,4S,5R,6S)-3,4,5-trihydroxy-6-((4-methyl-2-oxo-2H-chromen-7-yl)oxy)tetrahydro-2H-pyran-2-carboxylic acid dihydrate
Molecular Weight (g/mol) 352.29
Formula Weight 388.1
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.