missing translation for 'onlineSavingsMsg'
Learn More

4-Methylumbelliferyl-beta-D-glucuronide, 98%

Product Code. p-7025609
Click to view available options
Quantity:
50 mg
250 mg
Unit Size:
250mg
50mg
This item is not returnable. View return policy

Product Code. 11484687

Brand: Thermo Scientific Alfa Aesar B21190.MF

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 6160-80-1 | C16H16O9 | 352.295 g/mol

It is a fluorogenic substrate for measuring of β-glucoronidiase activity, for rapid sensitive identification of Escherichia coli. MUG is a substrate for glucuronidase. It is ideal for plant molecular study as a way to examine gene expression under various promoters. And while it works well for measuring activity in plant lysate and leaf disks, it can also work in whole plant assays.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
It is a fluorogenic substrate for measuring of ß-glucoronidiase activity, for rapid sensitive identification of Escherichia coli. MUG is a substrate for glucuronidase. It is ideal for plant molecular study as a way to examine gene expression under various promoters. And while it works well for measuring activity in plant lysate and leaf disks, it can also work in whole plant assays.

Solubility
Soluble in hot water.

Notes
Light sensitive. Store away from alkali metals bases and light. Keep the container tightly closed and place it in a cool, dry and well ventilated condition. Store in freezer at -20°C.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 6160-80-1
Molecular Formula C16H16O9
Molecular Weight (g/mol) 352.295
MDL Number MFCD00036772
InChI Key ARQXEQLMMNGFDU-JHZZJYKESA-N
Synonym 4-methylumbelliferyl-beta-d-glucuronide, 4-methylumbelliferyl glucuronide, 4-methylumbelliferone glucuronide, mug trihydrate, 4-methylumbelliferyl beta-glucuronide, 4-methyl-2-oxo-2h-1-benzopyran-7-yl beta-d-glucopyranosiduronic acid, 2s,3s,4s,5r,6s-3,4,5-trihydroxy-6-4-methyl-2-oxo-2h-chromen-7-yl oxy tetrahydro-2h-pyran-2-carboxylic acid, 2s,3s,4s,5r,6s-3,4,5-trihydroxy-6-4-methyl-2-oxo-chromen-7-yl oxy-tetrahydropyran-2-carboxylic acid, 2s,3s,4s,6s,5r-3,4,5-trihydroxy-6-4-methyl-2-oxochromen-7-yloxy-2h-3,4,5,6-tetrahydropyran-2-carboxylic acid, 2s,3s,4s,5r,6s-3,4,5-trihydroxy-6-4-methyl-2-oxochromen-7-yl oxy oxane-2-carboxylic acid
PubChem CID 91553
ChEBI CHEBI:1904
IUPAC Name (2S,3S,4S,5R,6S)-3,4,5-trihydroxy-6-(4-methyl-2-oxochromen-7-yl)oxyoxane-2-carboxylic acid
SMILES CC1=CC(=O)OC2=C1C=CC(=C2)OC3C(C(C(C(O3)C(=O)O)O)O)O

Specifications

Quantity 50 mg
Sensitivity Light sensitive
Solubility Information Soluble in hot water.
Optical Rotation −106° (c=0.1 in Water)
Formula Weight 352.3
Percent Purity 98%
Chemical Name or Material 4-Methylumbelliferyl-beta-D-glucuronide

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.