missing translation for 'onlineSavingsMsg'
Learn More

4-Methylumbelliferyl Beta-D-Glucoside, TRC

Product Code. 30133605
Change view
Click to view available options
Quantity:
1 g
5 g
10 g
Unit Size:
10g
1g
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30133605 5 g 5g
30134605 10 g 10g
17979344 1 g 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 30133605 Supplier Toronto Research Chemicals Supplier No. TRCM3345005G
 Restricted Item
Estimated Shipment: 02-12-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 18997-57-4
Molecular Formula C16 H18 O8
Molecular Weight (g/mol) 338.31
Synonym 2H-1-Benzopyran-2-one, 7-(β-D-glucopyranosyloxy)-4-methyl-, Coumarin, 7-(β-D-glucopyranosyloxy)-4-methyl- (6CI,8CI), 7-(β-D-Glucopyranosyloxy)-4-methyl-2H-1-benzopyran-2-one, 4-Methyl-coumarin-7-O-β-D-glucoside, 4-Methylumbelliferone β-D-glucoside, 4-Methylumbelliferyl glucoside, 4-Methylumbelliferyl β-D-glucopyranoside, 4-Methylumbelliferyl-β-D-glucoside
IUPAC Name 4-methyl-7-[(2S,3R,4S,5S,6R)-3,4,5-trihydroxy-6-(hydroxymethyl)oxan-2-yl]oxychromen-2-one
SMILES CC1=CC(=O)Oc2cc(O[C@@H]3O[C@H](CO)[C@@H](O)[C@H](O)[C@H]3O)ccc12

Specifications

Quantity 5 g
InChI Formula InChI=1S/C16H18O8/c1-7-4-12(18)23-10-5-8(2-3-9(7)10)22-16-15(21)14(20)13(19)11(6-17)24-16/h2-5,11,13-17,19-21H,6H2,1H3/t11-,13-,14+,15-,16-/m1/s1
Formula Weight 338.1002
Chemical Name or Material 4-Methylumbelliferyl-beta-D-glucopyranoside
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.