missing translation for 'onlineSavingsMsg'
Learn More

4-Methylumbelliferyl Alpha-L-Idopyranosiduronic Acid 2,4-Disulfate Trisodium Salt, TRC

Product Code. 30464516
Change view
Click to view available options
Quantity:
1 mg
10 mg
Unit Size:
10mg
1mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30464516 10 mg 10mg
30470071 1 mg 1mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 30464516 Supplier Toronto Research Chemicals Supplier No. TRCM33471010MG
 Restricted Item
Estimated Shipment: 25-08-2026
Add to basket
Add to basket
This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material 4-Methylumbelliferyl alpha-L-Idopyranosiduronic Acid 2,4-Disulfate Trisodium Salt
Molecular Formula C16H13Na3O15S2
Quantity 10 mg
Synonym 4-Methylumbelliferyl alpha-L-Iduronic Acid-2,4-disulfate Sodium Salt
InChI Formula InChI=1S/C16H16O15S2.3Na/c1-6-4-10(17)28-9-5-7(2-3-8(6)9)27-16-13(31-33(24,25)26)11(18)12(30-32(21,22)23)14(29-16)15(19)20;;;/h2-5,11-14,16,18H,1H3,(H,19,20)(H,21,22,23)(H,24,25,26);;;/q;3*+1/p-3/t11-,12-,13+,14+,16+;;;/m0.../s1
SMILES [Na+].[Na+].[Na+].CC1=CC(=O)Oc2cc(O[C@@H]3O[C@H]([C@@H](OS(=O)(=O)[O-])[C@H](O)[C@H]3OS(=O)(=O)[O-])C(=O)[O-])ccc12
Molecular Weight (g/mol) 578.36
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.