missing translation for 'onlineSavingsMsg'
Learn More

4-Methylumbelliferyl 2-Acetamido-2-deoxy-beta-D-glucopyranoside, TRC

Product Code. 30009202
Change view
Click to view available options
Quantity:
100 mg
1 g
5 g
Unit Size:
100mg
1g
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30009202 1 g 1g
30003397 5 g 5g
30005139 100 mg 100mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 30009202 Supplier Toronto Research Chemicals Supplier No. TRCM3340001G

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 37067-30-4
Molecular Formula C18 H21 N O8
Molecular Weight (g/mol) 379.36
Synonym 2H-1-Benzopyran-2-one, 7-[[2-(acetylamino)-2-deoxy-β-D-glucopyranosyl]oxy]-4-methyl-, Coumarin, 7-[(2-acetamido-2-deoxy-β-D-glucopyranosyl)oxy]-4-methyl- (7CI,8CI), 7-[[2-(Acetylamino)-2-deoxy-β-D-glucopyranosyl]oxy]-4-methyl-2H-1-benzopyran-2-one, 4-Methylumbelliferyl 2-acetamido-2-deoxy-β-D-glucopyranoside, 4-Methylumbelliferyl-N-acetyl-β-glucosaminide, 4-Methylumbelliferyl-N-acetylglucosaminide, 4-Methylumbelliferyl-β-D-N-acetylglucosaminide, 4-Methylumbelliferyl-β-N-acetylglucosaminide, Methylumbelliferyl 2-acetamido-2-deoxy-β-D-glucopyranoside
IUPAC Name N-[(2S,3R,4R,5S,6R)-4,5-dihydroxy-6-(hydroxymethyl)-2-(4-methyl-2-oxochromen-7-yl)oxyoxan-3-yl]acetamide
SMILES CC(=O)N[C@@H]1[C@@H](O)[C@H](O)[C@@H](CO)O[C@H]1Oc2ccc3C(=CC(=O)Oc3c2)C

Specifications

Quantity 1 g
InChI Formula InChI=1S/C18H21NO8/c1-8-5-14(22)26-12-6-10(3-4-11(8)12)25-18-15(19-9(2)21)17(24)16(23)13(7-20)27-18/h3-6,13,15-18,20,23-24H,7H2,1-2H3,(H,19,21)/t13-,15-,16-,17-,18-/m1/s1
Formula Weight 379.1267
Chemical Name or Material 4-Methylumbelliferyl 2-Acetamido-2-deoxy-beta-D-glucopyranoside
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.