missing translation for 'onlineSavingsMsg'
Learn More

4-Methylumbelliferone, TRC

Product Code. 30133039
Change view
Click to view available options
Quantity:
25 g
100 g
Unit Size:
100g
25g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30133039 100 g 100g
17962335 25 g 25g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 30133039 Supplier Toronto Research Chemicals Supplier No. TRCM333000100G
 Restricted Item
Estimated Shipment: 02-12-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 90-33-5
Molecular Formula C10 H8 O3
Molecular Weight (g/mol) 176.17
Synonym β-Methylumbelliferone, 4-MU, 4-Methyl-7-hydroxycoumarin, 4-Methylumbelliferone, 7-Hydroxy-4-methyl-2-chromenone, 7-Hydroxy-4-methyl-2-oxo-3-chromene, 7-Hydroxy-4-methyl-2H-1-benzopyran-2-one, 7-Hydroxy-4-methyl-2H-chromen-2-one, 7-Hydroxy-4-methylcoumarin, Bilcolic, Bilicante, Biliton H, Cantabilin, Cantabiline, Cholestil, Cholonerton, Cholspasmin, Coumarin 4, Coumarin 456, Cumarote C, Eurogale, Himecol, Hymecromone, Imecromone, LM 94, Medilla, Mendiaxon, NSC 19026, NSC 9408, Omega 127, Pilot 447
IUPAC Name 7-hydroxy-4-methylchromen-2-one
SMILES CC1=CC(=O)Oc2cc(O)ccc12

Specifications

Quantity 100 g
InChI Formula InChI=1S/C10H8O3/c1-6-4-10(12)13-9-5-7(11)2-3-8(6)9/h2-5,11H,1H3
Formula Weight 176.0473
Chemical Name or Material Hymecromone
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.