missing translation for 'onlineSavingsMsg'
Learn More

4-Methylumbelliferone, 97%

Product Code. 11424123
Change view
Click to view available options
Quantity:
50 g
250 g
1000 g
Unit Size:
1000g
250g
50g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11424123 250 g 250g
11414123 50 g 50g
11404123 1000 g 1000g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 11424123 Supplier Thermo Scientific Alfa Aesar Supplier No. A10337.30
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 90-33-5 | C10H8O3 | 176.17 g/mol

4-Methylumbelliferone is used in the determination of nitric acid and as a standard for fluorometric determination of enzyme activity. It is also used in bile therapy.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
4-Methylumbelliferone is used in the determination of nitric acid and as a standard for fluorometric determination of enzyme activity. It is also used in bile therapy.

Solubility
Soluble in alcohol. Insoluble in cold water. Soluble in acetic acid, alkali and ammonium hydroxide. Slightly soluble in hot water, chloroform and diethyl ether.

Notes
Store in a cool place. Incompatible with strong bases and strong oxidizing agents.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 90-33-5
Molecular Formula C10H8O3
Molecular Weight (g/mol) 176.17
MDL Number MFCD00006866
InChI Key HSHNITRMYYLLCV-UHFFFAOYSA-N
Synonym 4-methylumbelliferone, hymecromone, 7-hydroxy-4-methylcoumarin, imecromone, 7-hydroxy-4-methyl-2h-chromen-2-one, cholestil, mendiaxon, 4-methyl-7-hydroxycoumarin, beta-methylumbelliferone, bilcolic
PubChem CID 5280567
ChEBI CHEBI:17224
IUPAC Name 7-hydroxy-4-methylchromen-2-one
SMILES CC1=CC(=O)OC2=CC(O)=CC=C12

Specifications

Melting Point 186°C to 191°C
Quantity 250 g
Beilstein 142217
Merck Index 14,4854
Solubility Information Soluble in alcohol; Insoluble in cold water; Soluble in acetic acid,alkali and ammonium hydroxide; Slightly soluble in hot water,chloroform and diethyl ether.
Formula Weight 176.17
Percent Purity 97%
Chemical Name or Material 4-Methylumbelliferone
compliance-icons
Product Identifier
  • 4-Methylumbelliferone
Signal Word
  • Warning
Hazard Category
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H315-Causes skin irritation.
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P302+P352-IF ON SKIN: Wash with plenty of water/soap.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P312-Call a POISON CENTER/doctor/if you feel unwell.
  • P332+P313-If skin irritation occurs: Get medical advice/attention.
  • P337+P313-If eye irritation persists: Get medical advice/attention.
Supplemental information
  • MIXTURE LIST-Contain: 4-Methylumbelliferone

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.