missing translation for 'onlineSavingsMsg'
Learn More

Alfa Aesar™ 4-(Methylthio)thiophenol, 96%

Product Code. 11352147
Change view
Click to view available options
Quantity:
5 g
Unit Size:
5g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11352147 5 g 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 11352147 Supplier Alfa Aesar™ Supplier No. L09580.06

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

Chemical Identifiers

CAS 1122-97-0
Molecular Formula C7H8S2
Molecular Weight (g/mol) 156.261
MDL Number MFCD00082770
InChI Key KYMOWQQIZINTJZ-UHFFFAOYSA-N
Synonym 4-methylthio thiophenol, 4-methylthio benzenethiol, 4-methylthiothiophenol, 4-methylsulfanyl benzenethiol, 4-methylsulfanyl thiophenol, p-methylthiothiophenol, 4-mercaptothioanisole, pubchem6825, 4-methylthio-thiophenol
PubChem CID 136896
IUPAC Name 4-methylsulfanylbenzenethiol
SMILES CSC1=CC=C(C=C1)S

Specifications

Melting Point 12°C to 16°C
Boiling Point 116°C to 117°C (3 mmHg)
Refractive Index 1.653
Quantity 5 g
Beilstein 1931500
Sensitivity Air Sensitive
Formula Weight 156.27
Percent Purity 96%
Chemical Name or Material 4-(Methylthio)thiophenol
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.