missing translation for 'onlineSavingsMsg'
Learn More

4-(Methylthio)phenylzinc iodide, 0.5M in THF, packaged under Argon in resealable ChemSeal™ bottles, Thermo Scientific™

Product Code. 15405325
Change view
Click to view available options
Quantity:
50 mL
Packaging:
ChemSeal™ bottles
Unit Size:
50mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging unitSize
15405325 50 mL ChemSeal™ bottles 50mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 15405325 Supplier Thermo Scientific Alfa Aesar Supplier No. H58137.AD

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

  • Packaged under Argon
TRUSTED_SUSTAINABILITY

Specifications

CAS 307496-24-8
Color Black to Yellow
Packaging ChemSeal™ bottles
Molecular Formula C7H7ISZn
MDL Number MFCD01863641
Quantity 50 mL
UN Number UN3399
Sensitivity Air sensitive; May form a precipitate
Synonym 4-thiomethyl phenylzinc iodide, 4-methylthio phenylzinc iodide, iodo 4-methylsulfanyl phenyl zinc, 4-methylthio phenylzinc iodide solution, 0.5 m in thf, 4-thiomethyl phenylzinc iodide 0.5 m in tetrahydrofuran, 4-methylthio phenylzinc iodide, 0.5m in thf, packaged under argon in resealable chemseal bottles
InChI Key BSLXURHFHNFSCY-UHFFFAOYSA-M
SMILES [Zn+]I.CSC1=CC=[C-]C=C1
IUPAC Name iodozinc(1+);methylsulfanylbenzene
Molecular Weight (g/mol) 315.48
PubChem CID 24722603
Formula Weight 315.48
Physical Form Liquid
Chemical Name or Material 4-(Methylthio)phenylzinc iodide
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.