missing translation for 'onlineSavingsMsg'
Learn More

4-(Methylthio)phenylboronic acid, 97%, ACROS Organics™

Product Code. 10796442 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
1 g
5 g
25 g
Packaging:
Glass Bottle
Unit Size:
1g
25g
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging unitSize
10796442 1 g Glass Bottle 1g
10327191 5 g Glass Bottle 5g
10793255 25 g Glass Bottle 25g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 10796442 Supplier Thermo Scientific Acros Supplier No. 344680010
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

Chemical Identifiers

CAS 98546-51-1
Molecular Formula C7H9BO2S
Molecular Weight (g/mol) 168.017
MDL Number MFCD00093410
InChI Key IVUHTLFKBDDICS-UHFFFAOYSA-N
Synonym 4-methylthio phenylboronic acid, 4-thioanisoleboronic acid, 4-methylsulfanyl phenylboronic acid, 4-methylthiophenylboronic acid, 4-methylthio phenyl boronic acid, 4-methylsulfanylphenyl boronic acid, 4-methylthio phenylboronicacid, 4-methylthio benzeneboronic acid, thioanisole-4-boronic acid, 4-methylmercapto benzeneboronic acid
PubChem CID 2733959
IUPAC Name (4-methylsulfanylphenyl)boronic acid
SMILES B(C1=CC=C(C=C1)SC)(O)O

Specifications

CAS Min % 96.0
CAS Max % 100.0
Color White
Infrared Spectrum Authentic
Assay Percent Range 97%
Packaging Glass Bottle
Linear Formula CH3SC6H4B(OH)2
Quantity 1 g
Solubility Information Solubility in water: .. Other solubilities: soluble in methanol
Formula Weight 168.02
Percent Purity 97%
Physical Form Crystalline Powder
Chemical Name or Material 4-(Methylthio)phenylboronic acid
Show More Show Less
compliance-icons
Product Identifier
  • 4-(Methylthio)phenylboronic acid
Signal Word
  • Warning
Hazard Category
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
  • H315-Causes skin irritation.
Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P312-Call a POISON CENTER/doctor/if you feel unwell.
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.