missing translation for 'onlineSavingsMsg'
Learn More

4-(Methylthio)phenylacetic acid, 97%

Product Code. 11317089
Change view
Click to view available options
Quantity:
1 g
5 g
Unit Size:
1g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11317089 1 g 1g
11327089 5 g 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11317089 Supplier Thermo Scientific Alfa Aesar Supplier No. H26888.03
 Restricted Item
Estimated Shipment: 20-08-2026
Add to basket
This item is not returnable. View return policy

CAS: 16188-55-9 | C9H10O2S | 182.237 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 16188-55-9
Molecular Formula C9H10O2S
Molecular Weight (g/mol) 182.237
MDL Number MFCD00192325
InChI Key AHMLFHMRRBJCRM-UHFFFAOYSA-N
Synonym 4-methylthio phenylacetic acid, 4-methylthiophenylacetic acid, 2-4-methylthio phenyl acetic acid, 4-methylsulfanylphenyl acetic acid, benzeneacetic acid, 4-methylthio, 4-methylsulfanyl-phenyl-acetic acid, 2-4-methylsulfanyl phenyl acetic acid, 4-methylsulfanyl phenyl acetic acid, 2-4-methylthiophenyl acetic acid, 4-methylthiophenylaceticacid
PubChem CID 4983912
IUPAC Name 2-(4-methylsulfanylphenyl)acetic acid
SMILES CSC1=CC=C(C=C1)CC(=O)O

Specifications

Melting Point 98°C to 102°C
Quantity 1 g
Formula Weight 182.24
Percent Purity 97%
Chemical Name or Material 4-(Methylthio)phenylacetic acid

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.