missing translation for 'onlineSavingsMsg'
Learn More

4-(Methylthio)butanol, 99%

Product Code. 11485307
Change view
Click to view available options
Quantity:
1 g
5 g
25 g
Unit Size:
1g
25g
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11485307 25 g 25g
11465307 1 g 1g
11475307 5 g 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 11485307 Supplier Thermo Scientific Alfa Aesar Supplier No. B22673.14
 Restricted Item
In Stock
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

CAS: 20582-85-8 | C5H12OS | 120.21 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 20582-85-8
Molecular Formula C5H12OS
Molecular Weight (g/mol) 120.21
MDL Number MFCD00002973
InChI Key JNTVUHZXIJFHAU-UHFFFAOYSA-N
Synonym 4-methylthio-1-butanol, 4-methylthio butanol, 4-methylthiobutan-1-ol, unii-yl87ury6pc, 4-methylsulfanyl-1-butanol, 1-butanol, 4-methylthio, yl87ury6pc, 4-methylthio butan-1-ol, 4-methylsulfanyl butan-1-ol, 4-methylthio-1-butanol, polymer-bound
PubChem CID 519793
SMILES CSCCCCO

Specifications

Density 0.999
Boiling Point 95°C to 98°C (20 mmHg)
Flash Point 101°C (215°F)
Odor Unpleasant
Linear Formula CH3S(CH2)4OH
Refractive Index 1.487
Quantity 25 g
IUPAC Name 4-methylsulfanylbutan-1-ol
Formula Weight 120.21
Percent Purity 99%
Chemical Name or Material 4-(Methylthio)butanol
Show More Show Less

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.