missing translation for 'onlineSavingsMsg'
Learn More
Learn More
4-(Methylthio)benzenemethanol, TRC
Click to view available options
Quantity:
5 g
Unit Size:
5g
Specifications
Specifications
| Chemical Name or Material | 4-(Methylthio)benzenemethanol |
| CAS | 3446-90-0 |
| Molecular Formula | C26 H33 F O8 |
| Quantity | 5 g |
| Synonym | 4-(Hydroxymethyl)phenyl methyl Sulfide, 4-(Methylmercapto)benzyl alcohol, 4-(Methylthio)benzyl alcohol, 4-Methylsulfanylbenzyl alcohol, [4-(Methylsulfanyl)phenyl]methanol, p-(Methylthio)benzyl alcohol |
| InChI Formula | InChI=1S/C26H33FO8/c1-13(28)33-12-20(32)26-21(34-22(2,3)35-26)10-15-16-9-18(30)17-8-14(29)6-7-23(17,4)25(16,27)19(31)11-24(15,26)5/h6-8,15-16,18-19,21,30-31H,9-12H2,1-5H3/t15-,16+,18-,19+,21+,23+,24+,25-,26+/m1/s1 |
| SMILES | CC(=O)OCC(=O)[C@]12OC(C)(C)O[C@H]1C[C@@H]3[C@@H]4C[C@@H](O)C5=CC(=O)C=C[C@]5(C)[C@]4(F)[C@@H](O)C[C@]23C |
| IUPAC Name | [2-[(1S,2R,4S,8R,9S,11S,12S,13S,19R)-12-fluoro-11,19-dihydroxy-6,6,9,13-tetramethyl-16-oxo-5,7-dioxapentacyclo[10.8.0.02,9.04,8.013,18]icosa-14,17-dien-8-yl]-2-oxoethyl] acetate |
| Molecular Weight (g/mol) | 492.534 |
| Formula Weight | 492.216 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction